2-Fluoro-3-methylpyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Fluoro-3-picoline

F0533-1G F0533-5G
TCIAF0533-1GEA 285 SEK
TCIAF0533-1G TCIAF0533-5G
2-Fluoro-3-methylpyridine ≥98.0%
2-Fluoro-3-methylpyridine
Formel: C₆H₆FN
MW: 111,12 g/mol
Kokpunkt: 151°C
Flampunkt: 48°C
MDL Number: MFCD00041223
CAS nummer: 2369-18-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6064a554c451ad7409e92ac21303bff02ca60ba&vid=e6064a554c451ad7409e92ac21303bff02ca60ba&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR