Diphenyl phthalate ≥98.0%
Leverantör: TCI
Varning
Synonyms: Phthalic acid diphenyl ester, Phenyl phthalate
P0305-25G P0305-500G
TCIAP0305-25GEA 285 SEK
TCIAP0305-25G TCIAP0305-500G
Diphenyl phthalate ≥98.0%
Diphenyl phthalate
Formel: C₂₀H₁₄O₄
MW: 318.33 g/mol
Kokpunkt: 257°C
MDL Number: MFCD00003038
CAS nummer: 84-62-8
EINECS: 201-546-4
Merck Index: 14,07306
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58c74c4b69ec616b54d360eb0d513754fdf7e943&vid=58c74c4b69ec616b54d360eb0d513754fdf7e943&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C
Solubility in hot Methanol almost transparency