4-Aminobutyric acid ≥99.0%
Leverantör: TCI EUROPE
Synonyms: GABA, γ-Aminobutyric acid
A0282-25G A0282-500G A0282-100G
TCIAA0282-25GEA 230 SEK
TCIAA0282-25G TCIAA0282-500G TCIAA0282-100G
4-Aminobutyric acid ≥99.0%
4-Aminobutyric acid
Formel: C₄H₉NO₂
MW: 103,12 g/mol
Kokpunkt: 248 °C (1013 hPa)
Smältpunkt: 202 °C
Densitet: 1,21 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008226
CAS nummer: 56-12-2
EINECS: 200-258-6
Merck Index: 14,00430
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ed10fbfb285efa2f0e0517a13ab3442baf4bbaa&vid=1ed10fbfb285efa2f0e0517a13ab3442baf4bbaa&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White powder to crystaline
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in Water almost transparency