Sodium butyrate ≥98.0%
Leverantör: TCI
Synonyms:
Butyric acid sodium salt, Sodium Butyrate
S0519-100G
S0519-25G
TCIAS0519-100GEA
445
SEK
TCIAS0519-100G
TCIAS0519-25G
Sodium butyrate ≥98.0%
Sodium butyrate
Formel:
C₄H₇NaO₂ MW: 110.09 g/mol Smältpunkt: 253°C |
MDL Number:
MFCD00002816 CAS nummer: 156-54-7 EINECS: 205-857-6 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on dried substance) |
Drying loss | max. 5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...