2-Butynoic acid ethyl ester ≥98.0%
Leverantör: TCI
Varning
Synonyms: Ethyl 2-butynoate
T1052-1ML T1052-5ML
TCIAT1052-1MLEA 255 SEK
TCIAT1052-1ML TCIAT1052-5ML
2-Butynoic acid ethyl ester ≥98.0%
2-Butynoic acid ethyl ester
Formel: C₆H₈O₂
MW: 112.13 g/mol
Kokpunkt: 163°C
Flampunkt: 62°C
MDL Number: MFCD00015182
CAS nummer: 4341-76-8
EINECS: 224-395-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcd29634494eb73766ea4eaf3138dd9f0360d6c3&vid=dcd29634494eb73766ea4eaf3138dd9f0360d6c3&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %