etyloleat ≥70%
Leverantör: TCI EUROPE
Synonyms:
Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester
O0054-25ML
O0054-500ML
TCIAO0054-25MLEA
295
SEK
TCIAO0054-25ML
TCIAO0054-500ML
etyloleat ≥70%
etyloleat
Formel:
C₂₀H₃₈O₂ MW: 310,52 g/mol Kokpunkt: 209°C Smältpunkt: –32 °C Densitet: 0,87 g/cm³ (20 °C) Flampunkt: 181°C Storage Temperature: Omgivande |
MDL Number:
MFCD00009579 CAS nummer: 111-62-6 EINECS: 203-889-5 Merck Index: 14,06828 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 70.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...