Benzamid ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Benzoic acid amide

B0012-25G B0012-500G
TCIAB0012-25GEA 405 SEK
TCIAB0012-25G TCIAB0012-500G
Benzamid ≥99.0%
Benzamid
Formel: C₇H₇NO
MW: 121,14 g/mol
Kokpunkt: 288 °C (1013 hPa)
Smältpunkt: 124 °C
Densitet: 1,35 g/cm³ (20 °C)
Flampunkt: 180 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007968
CAS nummer: 55-21-0
EINECS: 200-227-7
Merck Index: 14,01060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2fcec5f7c478028c8f24551a9d0aff7d89d7b43&vid=a2fcec5f7c478028c8f24551a9d0aff7d89d7b43&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 99.0 %
Melting point 127.0 to 130.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR