Citraconic anhydride ≥98.0%

Leverantör: TCI
Fara

Synonyms: 3-Methylfuran-2,5-dione

M0365-25G M0365-500G M0365-100G
TCIAM0365-25GEA 215 SEK
TCIAM0365-25G TCIAM0365-500G TCIAM0365-100G
Citraconic anhydride ≥98.0%
Citraconic anhydride
Formel: C₅H₄O₃
Kokpunkt: 215°C
Smältpunkt: 7…8 °C
Densitet: 1,243 g/cm³ (20 °C)
Flampunkt: 101°C
Storage Temperature: Kylskåp
MDL Number: MFCD00005522
CAS nummer: 616-02-4
EINECS: 210-459-0
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6fbb1b6f3db8ef8f4dc7750915c7a57c811283f&vid=a6fbb1b6f3db8ef8f4dc7750915c7a57c811283f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Methanolysis Method) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR