Oxalylklorid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: Oxalyl chloride
O0082-25G O0082-500G O0082-100G
TCIAO0082-25GEA 245 SEK
TCIAO0082-25G TCIAO0082-500G TCIAO0082-100G
Oxalylklorid ≥98.0% (by GC, titration analysis)
Oxalylklorid
Formel: C₂Cl₂O₂
MW: 126,93 g/mol
Kokpunkt: 64°C
Smältpunkt: –12 °C
Densitet: 1,48 g/cm³ (20 °C)
Flampunkt: ≥ 100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000704
CAS nummer: 79-37-8
EINECS: 201-200-2
UN: 2922
ADR: 8,II
Merck Index: 14,06914
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8a26c11444f8d677fef4be97df3bed497ce350b&vid=b8a26c11444f8d677fef4be97df3bed497ce350b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %