N-Hydroxysuccinimide 98,0 Titrimetrisk

Leverantör: TCI

Synonyms: 1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu

H0623-25G H0623-500G H0623-100G
TCIAH0623-25GEA 310 SEK
TCIAH0623-25G TCIAH0623-500G TCIAH0623-100G
N-Hydroxysuccinimide 98,0 Titrimetrisk
N-Hydroxysuccinimide
Formel: C₄H₅NO₃
MW: 115.09 g/mol
Kokpunkt: 363
Storage Temperature: Kylskåp
MDL Number: MFCD00005516
CAS nummer: 6066-82-6
EINECS: 228-001-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e1b5c7b9a8d5bb7074d39186e2df832062f28e8&vid=1e1b5c7b9a8d5bb7074d39186e2df832062f28e8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR