Succinimide ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Pyrrolidine-2,5-dione
S0108-25G S0108-500G
TCIAS0108-25GEA 215 SEK
TCIAS0108-25G TCIAS0108-500G
Succinimide ≥98.0%
Succinimide
Formel: C₄H₅NO₂
MW: 99,09 g/mol
Kokpunkt: 130°C
Smältpunkt: 121…124 °C
Densitet: 1,418 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00005495
CAS nummer: 123-56-8
EINECS: 204-635-6
Merck Index: 14,08871
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=249b7c0490d17949c68ffd63d33817be5b4174ad&vid=249b7c0490d17949c68ffd63d33817be5b4174ad&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 124.0 to 127.0 deg-C
Solubility in Water almost transparency