Natrium L(+)-askorbat ≥98.0%

Leverantör: TCI EUROPE

Synonyms: L (+)-Askorbinsyra natriumsalt

A0539-25G A0539-500G
TCIAA0539-25GEA 245 SEK
TCIAA0539-25G TCIAA0539-500G
Natrium L(+)-askorbat ≥98.0%
Natrium L(+)-askorbat
Formel: C₆H₇NaO₆
MW: 198,11 g/mol
Smältpunkt: 219 °C
Densitet: 1,799 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00082340
CAS nummer: 134-03-2
EINECS: 205-126-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67a0f6169f83c3f8d4d79739d158a6ccd9db52cc&vid=67a0f6169f83c3f8d4d79739d158a6ccd9db52cc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 98.0 %
Specific rotation [a]20/D +103 to +108 deg (C=10, H2O)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR