2-Hydroxybenzophenone ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: (2-Hydroxyphenyl)(phenyl)methanone, o-Hydroxybenzophenone

H0470-25G H0470-5G
TCIAH0470-25GEA 1650 SEK
TCIAH0470-25G TCIAH0470-5G
2-Hydroxybenzophenone ≥95.0%
2-Hydroxybenzophenone
Formel: C₁₃H₁₀O₂
MW: 198,22 g/mol
Kokpunkt: 148°C
Smältpunkt: 34…37 °C
Densitet: 1,03 g/cm³ (172 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00002216
CAS nummer: 117-99-7
EINECS: 204-226-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea6c5dc86ff9c4882466538b283dc8182f909d56&vid=ea6c5dc86ff9c4882466538b283dc8182f909d56&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 37.0 to 41.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR