6-Methoxy-1-indanone ≥98.0%
Leverantör: TCI EUROPE
M1535-25G M1535-5G
TCIAM1535-25GEA 925 SEK
TCIAM1535-25G TCIAM1535-5G
6-Methoxy-1-indanone ≥98.0%
6-Methoxy-1-indanone
Formel: C₁₀H₁₀O₂
MW: 162,19 g/mol
Kokpunkt: 270 °C (1013 hPa)
Smältpunkt: 108 °C
MDL Number: MFCD00021232
CAS nummer: 13623-25-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9170437cfa14c2999f5b410f452d188f1282771&vid=a9170437cfa14c2999f5b410f452d188f1282771&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 107.0 to 111.0 deg-C