2'-Nitroacetophenone ≥96.0%
Leverantör: TCI
Varning
Synonyms: o-Nitroacetophenone, 1-(2-Nitrophenyl)ethanone
N0110-25G N0110-5G
TCIAN0110-25GEA 805 SEK
TCIAN0110-25G TCIAN0110-5G
2'-Nitroacetophenone ≥96.0%
2'-Nitroacetophenone
Formel: C₈H₇NO₃
MW: 165.15 g/mol
Kokpunkt: 179°C
Smältpunkt: 23
Storage Temperature: Omgivande
MDL Number: MFCD00007144
CAS nummer: 577-59-3
EINECS: 209-414-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e05c451c8857def91f0ae9eda72ecc2294ac9eb6&vid=e05c451c8857def91f0ae9eda72ecc2294ac9eb6&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorles to Amber powder to lump to clear liquid
Purity (GC) min. 96.0 %
Freezing point 24.0 to 28.0 deg-C