2-Aminobenzophenone ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: o-Aminobenzophenone
A0788-25G A0788-500G
TCIAA0788-25GEA 710 SEK
TCIAA0788-25G TCIAA0788-500G
2-Aminobenzophenone ≥98.0% (by GC, titration analysis)
2-Aminobenzophenone
Formel: C₁₃H₁₁NO
MW: 197,24 g/mol
Kokpunkt: 384 °C (1013 hPa)
Smältpunkt: 105…106 °C
Densitet: 1,32 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007713
CAS nummer: 2835-77-0
EINECS: 220-613-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e98831d9d33751744d17835619237720398a51f&vid=2e98831d9d33751744d17835619237720398a51f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 103.0 to 108.0 deg-C
Solubility in hot Methanol almost transparency