2'-Aminoacetophenone
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-(2-Aminophenyl)ethanone, o-Aminoacetophenone
A0250-25G
A0250-5G
TCIAA0250-25GEA
685
SEK
TCIAA0250-25G
TCIAA0250-5G
2'-Aminoacetophenone
2'-Aminoacetophenone
Formel:
C₈H₉NO MW: 135,17 g/mol Kokpunkt: 252 °C (1013 hPa) Smältpunkt: 20 °C Densitet: 1,12 g/cm³ (20 °C) Storage Temperature: Kylskåp |
MDL Number:
MFCD00007717 CAS nummer: 551-93-9 EINECS: 209-002-8 Merck Index: 14,00413 |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...