2-Oxoglutarsyra ≥99.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
2-Ketoglutaric acid, A-Ketoglutaric acid, α-Ketoglutaric acid, 2-Oxopentanedioic acid
K0005-25G
K0005-500G
K0005-100G
TCIAK0005-25GEA
285
SEK
TCIAK0005-25G
TCIAK0005-500G
TCIAK0005-100G
2-Oxoglutarsyra ≥99.0%
2-Oxoglutarsyra
Formel:
HOOCCH₂CH₂COCOOH MW: 146,1 g/mol Kokpunkt: 323 °C (1013 hPa) Smältpunkt: 112…116 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00004165 CAS nummer: 328-50-7 EINECS: 206-330-3 Merck Index: 14,05303 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 113.0 to 117.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...