2-Acetylbenzoic acid ≥98.0%

Leverantör: TCI
Varning

A1075-25G A1075-5G
TCIAA1075-25GEA 1425 SEK
TCIAA1075-25G TCIAA1075-5G
2-Acetylbenzoic acid ≥98.0%
2-Acetylbenzoic acid
Formel: C₉H₈O₃
Kokpunkt: 110 - 112 °C (2 -rr)
Smältpunkt: 112…115 °C
MDL Number: MFCD00002475
CAS nummer: 577-56-0
EINECS: 209-413-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b76efa16eb5d58999060515f330f57c62b7efa05&vid=b76efa16eb5d58999060515f330f57c62b7efa05&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR