4-Hydroxyphthalonitrile ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3,4-Dicyanophenol

H0813-1G H0813-5G
TCIAH0813-1GEA 885 SEK
TCIAH0813-1G TCIAH0813-5G
4-Hydroxyphthalonitrile ≥97.0%
4-Hydroxyphthalonitrile
Formel: C₈H₄N₂O
MW: 144,13 g/mol
MDL Number: MFCD00191653
CAS nummer: 30757-50-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51dd5de3ded8a6940c3760509954488b526d7fbd&vid=51dd5de3ded8a6940c3760509954488b526d7fbd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 210.0 to 215.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR