3-Nitrophthalonitrile 98,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: 3-Nitrobenzene-1,2-dicarbonitrile, 3-Nitro-1,2-dicyanobenzene, 3-Nitro-1,2-benzenedicarbonitrile

N0566-25G N0566-5G
TCIAN0566-25GEA 1100 SEK
TCIAN0566-25G TCIAN0566-5G
3-Nitrophthalonitrile 98,0 GC_ANALYSMETOD
3-Nitrophthalonitrile
Formel: C₈H₃N₃O₂
MW: 173.13 g/mol
Smältpunkt: 160
MDL Number: MFCD00191558
CAS nummer: 51762-67-5
EINECS: 450-650-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a45af161efb3840a7faed97acde95f0b88e0869f&vid=a45af161efb3840a7faed97acde95f0b88e0869f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 162.0 to 165.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR