Phthalonitrile ≥99.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
1,2-Benzenedicarbonitrile, Benzene-1,2-dicarbonitrile, 1,2-Dicyanobenzene
P0404-25G
P0404-500G
P0404-100G
TCIAP0404-25GEA
215
SEK
TCIAP0404-25G
TCIAP0404-500G
TCIAP0404-100G
Phthalonitrile ≥99.0%
Phthalonitrile
Formel:
C₈H₄N₂ MW: 128,13 g/mol Kokpunkt: 305 °C (1013 hPa) Smältpunkt: 138…141 °C Densitet: 1,1 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00001771 CAS nummer: 91-15-6 EINECS: 202-044-8 UN: 3439 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 140.0 to 143.0 deg-C |
Solubility in hot Methanol | almost transparency |
Ignition residue (Sulfate) | max. 0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...