Pyridine-2-carbonitrile ≥99.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 2-Cyanopyridine, 2-Pyridinecarbonitrile, Picolinonitrile

C0455-25G C0455-500G
TCIAC0455-25GEA 230 SEK
TCIAC0455-25G TCIAC0455-500G
Pyridine-2-carbonitrile ≥99.0%
Pyridine-2-carbonitrile
Formel: C₆H₄N₂
MW: 104,11 g/mol
Kokpunkt: 225°C
Smältpunkt: 26…28 °C
Densitet: 1,081 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006218
CAS nummer: 100-70-9
EINECS: 202-880-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50f12477d645d64a1527dc7d983ef822a026e5aa&vid=50f12477d645d64a1527dc7d983ef822a026e5aa&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR