4-Phenylbenzonitrile ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 4-Biphenylcarbonitrile, 4-Cyanobiphenyl, p-Phenylbenzonitrile, [1,1'-Biphenyl]-4-carbonitrile
C1299-25G C1299-5G
TCIAC1299-25GEA 1825 SEK
TCIAC1299-25G TCIAC1299-5G
4-Phenylbenzonitrile ≥98.0%
4-Phenylbenzonitrile
Formel: C₁₃H₉N
MW: 179,22 g/mol
Kokpunkt: 193°C
Storage Temperature: Omgivande
MDL Number: MFCD00001821
CAS nummer: 2920-38-9
EINECS: 220-860-2
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8c83a9d3d210cb43ead9a21c2999fde18c0de51&vid=e8c83a9d3d210cb43ead9a21c2999fde18c0de51&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 88.0 deg-C