2-Chloro-3-hydroxypyridine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 2-Chloro-3-pyridinol
C1152-10G C1152-25G
TCIAC1152-10GEA 470 SEK
TCIAC1152-10G TCIAC1152-25G
2-Chloro-3-hydroxypyridine ≥98.0% (by GC, titration analysis)
2-Chloro-3-hydroxypyridine
Formel: C₅H₄ClNO
MW: 129,55 g/mol
Kokpunkt: 212 °C (1013 hPa)
Smältpunkt: 170…171 °C
MDL Number: MFCD00006235
CAS nummer: 6636-78-8
EINECS: 229-635-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ea5fc9371ae7ac50d80ddc5829cb8718e313c81&vid=0ea5fc9371ae7ac50d80ddc5829cb8718e313c81&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 167.0 to 172.0 deg-C