Chlorosulphonyl isocyanate ≥98.0%
Leverantör: TCI
|
Fara
|
C0886-25G
C0886-500G
C0886-100G
TCIAC0886-25GEA
375
SEK
TCIAC0886-25G
TCIAC0886-500G
TCIAC0886-100G
Chlorosulphonyl isocyanate ≥98.0%
Chlorosulphonyl isocyanate
Formel:
CClNO₃S Kokpunkt: 107°C Smältpunkt: -80°C Densitet: 1,62 g/cm³ (20 °C) Storage Temperature: Kylskåp |
MDL Number:
MFCD00011608 CAS nummer: 1189-71-5 EINECS: 214-715-2 UN: 3265 ADR: 8,II |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...