4-Chloro-2-iodoanisole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-chloro-2-iodo-1-methoxybenzene

C2298-25G C2298-5G
TCIAC2298-25GEA 2925 SEK
TCIAC2298-25G TCIAC2298-5G
4-Chloro-2-iodoanisole ≥98.0%
4-Chloro-2-iodoanisole
Formel: C₇H₆ClIO
MW: 268,48 g/mol
MDL Number: MFCD00052858
CAS nummer: 52807-27-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64117cdf0a67610b458bf1bdfbad8451d6128727&vid=64117cdf0a67610b458bf1bdfbad8451d6128727&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 49.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR