2,4-Dichloro-5-fluoropyrimidine ≥98.0%

Leverantör: TCI
Varning

Synonyms: 5-Fluoro-2,4-dichloropyrimidine

D3374-25G D3374-5G
TCIAD3374-25GEA 1950 SEK
TCIAD3374-25G TCIAD3374-5G
2,4-Dichloro-5-fluoropyrimidine ≥98.0%
2,4-Dichloro-5-fluoropyrimidine
Formel: C₄HCl₂FN₂
Kokpunkt: 80°C
Storage Temperature: Kylskåp
MDL Number: MFCD00233551
CAS nummer: 2927-71-1
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b14843fbcbaf465eb1a83c02296d65d948b79ddf&vid=b14843fbcbaf465eb1a83c02296d65d948b79ddf&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR