2-Chloro-3-fluoropyridine ≥98.0%

Leverantör: TCI
Fara

C2447-25G C2447-5G
TCIAC2447-25GEA 2200 SEK
TCIAC2447-25G TCIAC2447-5G
2-Chloro-3-fluoropyridine ≥98.0%
2-Chloro-3-fluoropyridine
Formel: C₅H₃ClFN
Kokpunkt: 80°C
Flampunkt: 63°C
MDL Number: MFCD03095302
CAS nummer: 17282-04-1
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c83e6f41e3db0aa33dba382f318a82442dd6008&vid=2c83e6f41e3db0aa33dba382f318a82442dd6008&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR