Triklorisocyanursyra ≥95.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Trichloro-1,3,5-triazinetrion, 1,3,5-Trichlorohexahydro-1,3,5-triazine-2,4,6-trione, Trichlorocyanuric acid, TCCA, 1,3,5-Trichloro-1,3,5-triazine-2,4,6-trione, Trichloroisocyanuric acid
T0620-25G
T0620-500G
TCIAT0620-25GEA
215
SEK
TCIAT0620-25G
TCIAT0620-500G
Triklorisocyanursyra ≥95.0%
Triklorisocyanursyra
Formel:
C₃Cl₃N₃O₃ MW: 232,41 g/mol Kokpunkt: 413 °C (1013 hPa) Smältpunkt: 225…230 °C Densitet: 2,07 g/cm³ (20 °C) Flampunkt: ≥ 225 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00006553 CAS nummer: 87-90-1 EINECS: 201-782-8 UN: 2468 ADR: 5.1,II Merck Index: 14,09641 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Iodometric Titration) | min. 95.0 % |
Solubility in Acetone | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...