3,6-Dichloropyridazine ≥98.0%
Leverantör: TCI
|
Varning
|
D1465-25G
D1465-500G
TCIAD1465-25GEA
230
SEK
TCIAD1465-25G
TCIAD1465-500G
3,6-Dichloropyridazine ≥98.0%
3,6-Dichloropyridazine
Formel:
C₄H₂Cl₂N₂ Kokpunkt: 89 - 91 °C (0,2 -rr) Smältpunkt: 70 °C Densitet: 1,7 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00006466 CAS nummer: 141-30-0 EINECS: 205-478-6 ADR: 6.1,III |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 67.0 to 70.0 deg-C |
Solubility in Methanol | almost transparency |