Chlorotriphenylsilane ≥95.0%
Leverantör: TCI
|
Fara
|
T0939-25G
T0939-5G
TCIAT0939-25GEA
885
SEK
TCIAT0939-25G
TCIAT0939-5G
Chlorotriphenylsilane ≥95.0%
Chlorotriphenylsilane
Formel:
C₁₈H₁₅ClSi Kokpunkt: 378 °C (1013 hPa) Smältpunkt: 90…95 °C Densitet: 1,0165 g/cm³ (20 °C) Storage Temperature: Kylskåp |
MDL Number:
MFCD00000496 CAS nummer: 76-86-8 EINECS: 200-989-0 UN: 3261 ADR: 8,II |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 93.0 to 98.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...