Chlorodimethylsilane ≥95.0%

Leverantör: TCI
Fara

Synonyms: Dimethylchlorosilane

C0778-25ML C0778-250ML
TCIAC0778-25MLEA 590 SEK
TCIAC0778-25ML TCIAC0778-250ML
Chlorodimethylsilane ≥95.0%
Chlorodimethylsilane
Formel: C₂H₇ClSi
MW: 94.62 g/mol
Kokpunkt: 37°C
Smältpunkt: –111°C
Flampunkt: –20°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000495
CAS nummer: 1066-35-9
EINECS: 213-912-0
UN: 2985
REACH: 01-2119486782-25
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02a02034b22ac17ef74eda6e6a0200db17f3aac1&vid=02a02034b22ac17ef74eda6e6a0200db17f3aac1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR