2,3,5-Trikloropyridin ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 2,3,5-TCP
T1479-25G T1479-250G
TCIAT1479-25GEA 350 SEK
TCIAT1479-25G TCIAT1479-250G
2,3,5-Trikloropyridin ≥98.0%
2,3,5-Trikloropyridin
Formel: C₅H₂Cl₃N
MW: 182,44 g/mol
Kokpunkt: 220°C
MDL Number: MFCD00043007
CAS nummer: 16063-70-0
EINECS: 407-270-2
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcb7e9cdb83095f7782c36042061d101d8717298&vid=dcb7e9cdb83095f7782c36042061d101d8717298&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 49.0 deg-C