α,4-Dichloro-α-phenyltoluene ≥96.0%

Leverantör: TCI
Fara

Synonyms: 4-Chlorobenzhydryl chloride, 1-Chloro-4-(chloro(phenyl)methyl)benzene

C1234-25G C1234-500G
TCIAC1234-25GEA 375 SEK
TCIAC1234-25G TCIAC1234-500G
α,4-Dichloro-α-phenyltoluene ≥96.0%
α,4-Dichloro-α-phenyltoluene
Formel: C₁₃H₁₀Cl₂
Kokpunkt: 160°C
Densitet: 1,24 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000856
CAS nummer: 134-83-8
EINECS: 205-158-6
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b797b56134134bba9d7ceedb81e2d4ab23fe3b0&vid=9b797b56134134bba9d7ceedb81e2d4ab23fe3b0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR