2-Bromo-3-hydroxypyridine ≥98.0%
Leverantör: TCI
Varning
B1478-25G B1478-5G
TCIAB1478-25GEA 1325 SEK
TCIAB1478-25G TCIAB1478-5G
2-Bromo-3-hydroxypyridine ≥98.0%
2-Bromo-3-hydroxypyridine
Formel: C₅H₄BrNO
Kokpunkt: 232 °C (1013 hPa)
Smältpunkt: 185…188 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006220
CAS nummer: 6602-32-0
EINECS: 229-547-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07f669e251e00727527f1fc46022e1cb9224ac3a&vid=07f669e251e00727527f1fc46022e1cb9224ac3a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Gray to Brown powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 179.0 to 188.0 deg-C