(3-Bromopropyl)trimethoxysilane ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Trimethoxy(3-bromopropyl)silane, 3-(Trimethoxysilyl)propylbromide

B3289-5G B3289-25G
TCIAB3289-5GEA 1150 SEK
TCIAB3289-5G TCIAB3289-25G
(3-Bromopropyl)trimethoxysilane ≥98.0%
(3-Bromopropyl)trimethoxysilane
Formel: BrH₂CCH₂CH₂Si(OCH₃)₃
MW: 243,17 g/mol
Flampunkt: 82°C
MDL Number: MFCD00053894
CAS nummer: 51826-90-5
EINECS: 257-453-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0195eda0b83bcec1b789e8bbe4b7fdef874df29d&vid=0195eda0b83bcec1b789e8bbe4b7fdef874df29d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR