α-Bromo-2-iodotoulene ≥98.0%

Leverantör: TCI
Fara

Synonyms: α-Bromo-o-iodotoulene, 2-Iodobenzyl bromide, 1-(Bromomethyl)-2-iodobenzene

I0635-5G I0635-1G
TCIAI0635-5GEA 750 SEK
TCIAI0635-5G TCIAI0635-1G
α-Bromo-2-iodotoulene ≥98.0%
α-Bromo-2-iodotoulene
Formel: C₇H₆BrI
MW: 296.93 g/mol
Kokpunkt: 125°C
Smältpunkt: 55.5
MDL Number: MFCD00236046
CAS nummer: 40400-13-3
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03552f1b29bbf31ce2ed021bffda52fd28c91b82&vid=03552f1b29bbf31ce2ed021bffda52fd28c91b82&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR