11-Bromoundecanoic acid ≥97.0% (by GC, titration analysis)
Leverantör: TCI
Varning
B0389-25G B0389-500G B0389-100G
TCIAB0389-25GEA 535 SEK
TCIAB0389-25G TCIAB0389-500G TCIAB0389-100G
11-Bromoundecanoic acid ≥97.0% (by GC, titration analysis)
11-Bromoundecanoic acid
Formel: BrH₂C(CH₂)₉COOH
Kokpunkt: 190°C
Smältpunkt: 48…49 °C
Densitet: 1,24 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002732
CAS nummer: 2834-05-1
EINECS: 220-602-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7238cd6fde65b454bf44b0e59ba2f8ee6f62d4e&vid=f7238cd6fde65b454bf44b0e59ba2f8ee6f62d4e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 48.0 to 52.0 deg-C
Solubility in Methanol almost transparency