2-Bromo-4,6-dichloroaniline ≥97.0%
Leverantör: TCI EUROPE
Varning
B2197-25G B2197-5G
TCIAB2197-25GEA 2100 SEK
TCIAB2197-25G TCIAB2197-5G
2-Bromo-4,6-dichloroaniline ≥97.0%
2-Bromo-4,6-dichloroaniline
Formel: C₆H₄BrCl₂N
MW: 240,91 g/mol
Kokpunkt: 273°C
Smältpunkt: 77…78 °C
MDL Number: MFCD00040936
CAS nummer: 697-86-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8108fdde34cfe44ce5343ed8e485402cd005ef49&vid=8108fdde34cfe44ce5343ed8e485402cd005ef49&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 80.0 to 84.0 deg-C