Flasks, flat bottom with standard ground joint
Leverantör: GLASWARENFABRIK KARL HECHT
41911030 41911036 41911006 41911012 41911018 41911024
HECH41911030EA 915 SEK
HECH41911030 HECH41911036 HECH41911006 HECH41911012 HECH41911018 HECH41911024
Flasks, flat bottom with standard ground joint
Kolvar Kolv, plan botten
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.