1-Methyl-1H-1,2,4-triazole ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Methyl-1,2,4-triazole
L18337.03 L18337.06
L18337.03EA 1475 SEK
L18337.03 L18337.06
1-Methyl-1H-1,2,4-triazole ≥99%
1-Methyl-1H-1,2,4-triazole
Purity: 99% CAS No.: 6086-21-1 Molecular Formula: C3H5N3 Formula Weight: 83.10 Flash Point: 81°(177°F) Refractive Index: 1.4705 Melting Point: 16° Boiling Point: 177-178° Beilstein Reference: 106245 MDL No.: MFCD01076192
Formel: C₃H₅N₃
MW: 83,10 g/mol
Kokpunkt: 177…178 °C
Smältpunkt: 16 °C
Densitet: 1,10
Flampunkt: 81 °C (177 °F)
Storage Temperature: Omgivande
MDL Number: MFCD01076192
CAS nummer: 6086-21-1