2-Ethyl-3,5(or 6)-dimethylpyrazine (mixture of isomers) ≥99%
Leverantör: Thermo Scientific
Varning
H27383.14 H27383.06
H27383.14EA 3325 SEK
H27383.14 H27383.06
2-Ethyl-3,5(or 6)-dimethylpyrazine (mixture of isomers) ≥99%
2-Ethyl-3,5(or 6)-dimethylpyrazine (mixture of isomers)
Purity: 99% CAS No.: 13925-07-0 Molecular Formula: C8H12N2 Formula Weight: 136.19 Flash Point: 66°(151°F) Refractive Index: 1.5015 Boiling Point: 182° MDL No.: MFCD00047392
Formel: C₈H₁₂N₂
MW: 136,194 g/mol
Kokpunkt: 182 °C
Densitet: 0,96
Flampunkt: 66 °C (151 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00047392
CAS nummer: 27043-05-6
EINECS: 237-694-1