Ethyl-4-oxocyclohexanecarboxylate ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Ethyl 4-oxocyclohexanecarboxylate, 4-(Ethoxycarbonyl)-1-cyclohexanone
C1530-25G C1530-5G
TCIAC1530-25GEA 1925 SEK
TCIAC1530-25G TCIAC1530-5G
Ethyl-4-oxocyclohexanecarboxylate ≥98.0%
Ethyl-4-oxocyclohexanecarboxylate
Formel: C₉H₁₄O₃
MW: 170,21 g/mol
Kokpunkt: 152°C
MDL Number: MFCD00013285
CAS nummer: 17159-79-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a8f0ad7a79543401c8d1b766d430a27ecd5ff52&vid=8a8f0ad7a79543401c8d1b766d430a27ecd5ff52&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %