O-Benzylhydroxylamine hydrochloride 99%
Leverantör: Thermo Fisher Scientific
A17393.06 A17393.14 A17393.22
A17393.06EA 990 SEK
A17393.06 A17393.14 A17393.22
O-Benzylhydroxylamine hydrochloride 99%
O-Benzylhydroxylamine hydrochloride
Purity: 99% CAS No.: 2687-43-6 Molecular Formula: C6H5CH2ONH2.HCl Formula Weight: 159.62 Melting Point: ca 238° subl. Beilstein Reference: 3595440
Formel: C₇H₁₀ClNO
Smältpunkt: ∼ 238 °C (subl.)
Storage Temperature: Omgivande
MDL Number: MFCD00012952
CAS nummer: 2687-43-6
EINECS: 220-249-0