2,6-Xylyl isocyanide
Leverantör: Thermo Fisher Scientific
Fara
20748.03 20748.06
20748.03EA 2875 SEK
20748.03 20748.06
2,6-Xylyl isocyanide
2,6-Xylyl isocyanide
CAS No.: 2769-71-3 Molecular Formula: C9H9N Formula Weight: 131.18 Physical Form: Crystalline Odour: Unpleasant Melting Point: 73-74° UN No.: 2811 MDL No.: MFCD00013479
Formel: C₉H₉N
MW: 131.18 g/mol
Smältpunkt: 73…74 °C
Storage Temperature: Omgivande
MDL Number: MFCD00013479
CAS nummer: 2769-71-3
EINECS: 220-459-2
UN: 2811
ADR: 6.1 III