Mucobromic acid ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,3-Dibromomalealdehydic acid, (Z)-2,3-Dibromo-4-oxobut-2-enoic acid
A15998.22 A15998.36
A15998.22EA 2075 SEK
A15998.22 A15998.36
Mucobromic acid ≥98%
Mucobromic acid
Purity: 98+% CAS No.: 488-11-9 Molecular Formula: C4H2Br2O3 Formula Weight: 257.88 Melting Point: 121-124° Beilstein Reference: 1705707 UN No.: 3261 MDL No.: MFCD00063745
Formel: C₄H₂Br₂O₃
MW: 257,88 g/mol
Smältpunkt: 121…124 °C
Storage Temperature: Omgivande
MDL Number: MFCD00063745
CAS nummer: 488-11-9
EINECS: 207-670-5
UN: 3261
ADR: 8,III