Tetrabutylammonium acetate 1.0 M i vattenlösning
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Tetra-n-butylammonium acetate
B24773.14 B24773.22
B24773.14EA 1300 SEK
B24773.14 B24773.22
Tetrabutylammonium acetate 1.0 M i vattenlösning
Tetrabutylammonium acetate
CAS No.: 10534-59-5 Molecular Formula: C18H39NO2 Formula Weight: 301.51 Refractive Index: 1.3890 Boiling Point: 100° Beilstein Reference: 3599376
Formel: C₁₈H₃₉NO₂
MW: 301.51 g/mol
Kokpunkt: 100 °C
Densitet: 0.990
Storage Temperature: Omgivande
MDL Number: MFCD00043208
CAS nummer: 10534-59-5
EINECS: 234-101-8