Decyltrimethylammonium bromide ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (1-Decyl)trimethylammonium bromide, N,N,N-Trimethyldecan-1-aminium bromide
L12144.09 L12144.18
L12144.09EA 1075 SEK
L12144.09 L12144.18
Decyltrimethylammonium bromide ≥98%
Decyltrimethylammonium bromide
Purity: 98% CAS No.: 2082-84-0 Molecular Formula: CH3(CH2)9N(CH3)3Br Formula Weight: 280.30 Melting Point: 243-245° Beilstein Reference: 3915222
Formel: C₁₃H₃₀BrN
MW: 280.30 g/mol
Smältpunkt: 243…245 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041973
CAS nummer: 2082-84-0
EINECS: 218-219-7