1,4,7-Triazacyclononane trihydrochloride ≥98.0% (by total nitrogen and titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 1,4,7-Triazonane trihydrochloride

T1600-1G T1600-5G
TCIAT1600-1GEA 1675 SEK
TCIAT1600-1G TCIAT1600-5G
1,4,7-Triazacyclononane trihydrochloride ≥98.0% (by total nitrogen and titration analysis)
1,4,7-Triazacyclononane trihydrochloride
Formel: C₆H₁₈Cl₃N₃
MW: 238,5900 g/mol
MDL Number: MFCD00074887
CAS nummer: 58966-93-1
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e38e4850eb9672e8078df882b4ac99b9461aa36d&vid=e38e4850eb9672e8078df882b4ac99b9461aa36d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR